Please login to your online account to display your discounted pricing

4-Propoxyphenol, 96%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213320500

 View more versions of this product

Catalog No. AC213320500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.19
Quantity 50g
TSCA TSCA
CAS Min % 95.0
CAS Max % 100.0
Synonym Hydroquinone monopropyl ether
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4-Propoxyphenol
Molecular Formula C9H12O2
Assay Percent Range 96%
MDL Number MFCD00002335
CAS 18979-50-5
Name Note 96%
Linear Formula CH3CH2CH2OC6H4OH

Specifications

Chemical Name or Material 4-Propoxyphenol
Name Note 96%
CAS 18979-50-5
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C9H12O2
Linear Formula CH3CH2CH2OC6H4OH
Formula Weight 152.19
MDL Number MFCD00002335
Synonym Hydroquinone monopropyl ether
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents