Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Propionyloxybenzaldehyde, 95%

$104.53 - $397.14

Chemical Identifiers

Chemical Name or Material 4-Propionyloxybenzaldehyde
CAS 50262-48-1
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00043522
Beilstein 2966420
Synonym 4-Hydroxybenzaldehyde propionate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0873206

  SDS
alfa aesar™
L0873206
5g Each for $104.53

AAL0873214

  SDS
alfa aesar™
L0873214
25g Each for $397.14
Description & Specifications

Specifications

Formula Weight 178.19
Chemical Name or Material 4-Propionyloxybenzaldehyde
MDL Number MFCD00043522
CAS 50262-48-1
Beilstein 2966420
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
TSCA TSCA
Assay 95%
Molecular Formula C10H10O3
Synonym 4-Hydroxybenzaldehyde propionate
Boiling Point 132° to 136°C (0.5mmHg)
Flash Point >110°C (230°F)
Quantity 5g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.