Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Propargylaminocarbonyl)benzeneboronic acid pinacol ester, 95%

$88.20 - $338.64

Chemical Identifiers

Chemical Name or Material 4-(Propargylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1218790-49-8
Molecular Formula C16H20BNO3
Formula Weight 285.15
MDL Number MFCD11113041
Synonym N-(2-Propyn-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5112503

  SDS
alfa aesar™
H5112503
1g Each for $88.20

AAH5112506

  SDS
alfa aesar™
H5112506
5g Each for $338.64
Description & Specifications

Specifications

Formula Weight 285.15
Chemical Name or Material 4-(Propargylaminocarbonyl)benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C16H20BNO3
MDL Number MFCD11113041
CAS 1218790-49-8
Synonym N-(2-Propyn-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Quantity 1g