Please login to your online account to display your discounted pricing

4-PPBP maleate, Tocris


Specific σ ligand, NMDA antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS062050

 View more versions of this product

Catalog No. 062050

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-PPBP maleate
Quantity 50mg
CAS 201216-39-9
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C21H27N·C4H4O4
Formula Weight 409.52
Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate

Specifications

Chemical Name or Material 4-PPBP maleate
Assay Percent Range >99%
Molecular Formula C21H27N·C4H4O4
Formula Weight 409.52
Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate
Accessories

Store at Room Temperature