Please login to your online account to display your discounted pricing

4-PPBP maleate, Tocris

Specific σ ligand, NMDA antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material 4-PPBP maleate
Assay Percent Range >99%
Molecular Formula C21H27N·C4H4O4
Formula Weight 409.52
Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

062050

 
r & d systems
RDS062050
50mg Each for $697.40

062010

 
r & d systems
RDS062010
10mg Each for $168.15
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 409.52
Chemical Name or Material 4-PPBP maleate
CAS 201216-39-9
Assay Percent Range >99%
Molecular Formula C21H27N·C4H4O4
Synonym 4-Phenyl-1-(4-phenylbutyl)piperidine maleate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature