Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Piperidone hydrochloride monohydrate, 98%

$47.91 - $679.55

Chemical Identifiers

Chemical Name or Material 4-Piperidone hydrochloride monohydrate
CAS 40064-34-4
Molecular Formula C5H9NO·HCl·H2O
Formula Weight 153.61 (135.59 Anhydrous)
MDL Number MFCD00012776
Beilstein 3916558
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1372609

  SDS
alfa aesar™
A1372609
10g Each for $47.91

AAA1372618

  SDS
alfa aesar™
A1372618
50g Each for $158.22

AAA1372630

  SDS
alfa aesar™
A1372630
250g Each for $679.55
Description & Specifications

Specifications

Formula Weight 153.61 (135.59 Anhydrous)
Assay 98%
Molecular Formula C5H9NO·HCl·H2O
Quantity 10g
MDL Number MFCD00012776
Melting Point 94° to 100°C
CAS 40064-34-4
Chemical Name or Material 4-Piperidone hydrochloride monohydrate
Beilstein 3916558

Applications
4-Piperidone hydrochloride monohydrate is used in the generation of compound libraries based on sub-reactions of an Ugi multicomponent reaction (MCR).

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.