missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidone Ethyleneketal, TCI America™

Manufacturer:  TCI America P232525G

 View more versions of this product

Catalog No. P232525G



Specifications

Specifications

4-Piperidone Ethyleneketal
Yellow
177-11-7
MFCD00005976
KPKNTUUIEVXMOH-UHFFFAOYSA-N
1,4-dioxa-8-azaspiro[4.5]decane
67435
Clear Liquid at 20°C
110°C
25g
C7H13NO2
1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4.5 decane, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
C1CNCCC12OCCO2
143.186
143.19
≥98.0% (GC,T)

Chemical Identifiers

177-11-7
143.186
KPKNTUUIEVXMOH-UHFFFAOYSA-N
67435
C1CNCCC12OCCO2
C7H13NO2
MFCD00005976
1,4-dioxa-8-azaspiro 4.5 decane, 4-piperidone ethylene ketal, 4-piperidinone ethyl ketal, 4-piperidone-ethylene ketal, 4-piperidone ethylene acetal, 1,4-dioxa-8-azaspiro 4.5 decane, 1,4-dioxa-8-azaspiro 4,5 decane, 4-piperidoneethyleneketal, piperidone-4-ethyleneketal, 1,4-dioxa-8-aza-spiro 4.5 decane
1,4-dioxa-8-azaspiro[4.5]decane
Safety and Handling

Safety and Handling

TSCA : No

Documents