Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Piperidone ethylene ketal, 98+%

$76.54 - $1,054.73

Chemical Identifiers

Chemical Name or Material 4-Piperidone ethylene ketal
CAS 177-11-7
Molecular Formula C7H13NO2
Formula Weight 143.19
MDL Number MFCD00005976
Beilstein 506799
Synonym 1,4-Dioxa-8-azaspiro[4.5]decane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1241909

  SDS
alfa aesar™
A1241909
10g Each for $76.54

AAA1241918

  SDS
alfa aesar™
A1241918
50g Each for $254.65

AAA1241930

  SDS
alfa aesar™
A1241930
250g Each for $1,054.73
Description & Specifications

Specifications

Formula Weight 143.19
MDL Number MFCD00005976
CAS 177-11-7
Molecular Formula C7H13NO2
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1,4-Dioxa-8-azaspiro[4.5]decane
Quantity 10g
Health Hazard 1 Warning
Boiling Point 99° to 101°C (14mmHg)
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material 4-Piperidone ethylene ketal
Assay ≥98%
Flash Point 81°C (177°F)
Beilstein 506799

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.