missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Piperidone ethylene ketal, 98+%

Manufacturer:  Alfa Aesar™ A1241930

 View more versions of this product

Catalog No. AAA1241930


 

Description

Description

Applications
4-Piperidone ethylene ketal is used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes.

Solubility
Soluble in water (Partly miscible).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Specifications

Specifications

4-Piperidone ethylene ketal
99° to 101°C (14mmHg)
177-11-7
C7H13NO2
506799
KPKNTUUIEVXMOH-UHFFFAOYSA-N
1,4-dioxa-8-azaspiro[4.5]decane
67435
81°C (177°F)
250g
≥98%
MFCD00005976
1,4-Dioxa-8-azaspiro[4.5]decane
C1CNCCC12OCCO2
143.186
143.19

Chemical Identifiers

177-11-7
143.186
KPKNTUUIEVXMOH-UHFFFAOYSA-N
67435
C7H13NO2
MFCD00005976
1,4-Dioxa-8-azaspiro[4.5]decane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents