Please login to your online account to display your discounted pricing

4-Piperidinyl(1-pyrrolidinyl)methanone, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO01169DA

 View more versions of this product

Catalog No. MO01169DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 182.27
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Molecular Formula C10H18N2O
Assay Percent Range 97%
CAS 35090-95-0
Health Hazard 2 GHS P Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Synonym 4-Piperidinyl(1-pyrrolidinyl)methanone (IUPAC)
Chemical Name or Material 4-Piperidinyl(1-pyrrolidinyl)methanone
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03372520

Specifications

Chemical Name or Material 4-Piperidinyl(1-pyrrolidinyl)methanone
CAS 35090-95-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H18N2O
Formula Weight 182.27
MDL Number MFCD03372520
Synonym 4-Piperidinyl(1-pyrrolidinyl)methanone (IUPAC)
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents