missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidinyl(1-pyrrolidinyl)methanone, 97%, Maybridge

$137.44 - $137.44

Chemical Identifiers

CAS 35090-95-0
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C10H18N2O
Formula Weight 182.27
MDL Number MFCD03372520
Synonym 4-Piperidinyl(1-pyrrolidinyl)methanone (IUPAC)
Chemical Name or Material 4-Piperidinyl(1-pyrrolidinyl)methanone
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO01169DA SDS Fisher Scientific Company
MO01169DA
Amber Glass Bottle 1g Each for $137.44
Specifications Description & Specifications

Chemical Identifiers

35090-95-0
97%
182.27
4-Piperidinyl(1-pyrrolidinyl)methanone (IUPAC)
97%
97.0
C10H18N2O
MFCD03372520
4-Piperidinyl(1-pyrrolidinyl)methanone

Specifications

182.27
97.0
C10H18N2O
35090-95-0
MFCD03372520
97%
1g
Amber Glass Bottle
97%
4-Piperidinyl(1-pyrrolidinyl)methanone (IUPAC)
4-Piperidinyl(1-pyrrolidinyl)methanone

Safety and Handling

GHS P Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger