Please login to your online account to display your discounted pricing

4-(Piperidinosulfonyl)aniline, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00817EA

 View more versions of this product

Catalog No. TL00817EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 240.33
Quantity 10g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Assay Percent Range 97%
CAS 6336-68-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Chemical Name or Material 4-(Piperidinosulfonyl)aniline
MDL Number MFCD00276969
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C11H16N2O2S
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-(Piperidinosulfonyl)aniline
CAS 6336-68-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H16N2O2S
Formula Weight 240.33
MDL Number MFCD00276969
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.

SDS & Safety Documents