missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidinopiperidine, TCI America™

Manufacturer:  TCI America P163825G

 View more versions of this product

Catalog No. P163825G



Specifications

Specifications

4-Piperidinopiperidine
66°C
4897-50-1
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
C1CCN(CC1)C2CCNCC2
168.284
CHEBI:40117
Crystal-Powder at 20°C
White-Yellow
25g
C10H20N2
QDVBKXJMLILLLB-UHFFFAOYSA-N
1-piperidin-4-ylpiperidine
78607
168.28
≥98.0% (GC)

Chemical Identifiers

4897-50-1
168.284
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
CHEBI:40117
C1CCN(CC1)C2CCNCC2
C10H20N2
QDVBKXJMLILLLB-UHFFFAOYSA-N
78607
1-piperidin-4-ylpiperidine
Safety and Handling

Safety and Handling

TSCA : No

Documents