missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidinopiperidine, 95%, Tech., ACROS Organics™

Manufacturer:  Acros Organics 165450500

 View more versions of this product

Catalog No. AC165450500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Piperidinopiperidine
Glass bottle
62.0°C to 67.0°C
tech., 95%
94.0
95%
MFCD00006475
QDVBKXJMLILLLB-UHFFFAOYSA-N
1-piperidin-4-ylpiperidine
78607
168.28
95%
Authentic
Brown-Yellow to Yellow
50g
4897-50-1
100.0
C10H20N2
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
C1CCN(CC1)C2CCNCC2
168.284
CHEBI:40117
Crystalline Solid
Technical

Chemical Identifiers

4897-50-1
168.284
QDVBKXJMLILLLB-UHFFFAOYSA-N
78607
1-piperidin-4-ylpiperidine
C10H20N2
MFCD00006475
4-piperidinopiperidine, 1,4'-bipiperidine, 4-piperidino-piperidine, 1,4'-bipiperidyl, 4-1-piperidinyl piperidine, 1,4' bipiperidinyl, 4-piperidin-1-yl piperidine, 1,4-bipiperidine, 1-piperidin-4-yl piperidine, 4-piperidin-1-ylpiperidine
CHEBI:40117
C1CCN(CC1)C2CCNCC2
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 225-522-8

SDS Documents