Please login to your online account to display your discounted pricing

4-Piperidinopiperidine, 95%, Tech., ACROS Organics™

$227.33 - $227.33

Chemical Identifiers

Chemical Name or Material 4-Piperidinopiperidine
Name Note tech., 95%
CAS 4897-50-1
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C10H20N2
Formula Weight 168.28
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165450100

  SDS
thermo fisher scientific
165450100
Glass bottle 10g Each for $227.33

AC165450500

  SDS
fisher scientific acros
165450500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Crystalline Solid
Assay ≥94%
Formula Weight 168.28
Quantity 10g
Color Light yellow to yellow-brown
EINECS Number 225-522-8
Melting Point 62° to 67°C
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00006475
Packaging Glass bottle
Chemical Name or Material 4-Piperidinopiperidine
Name Note tech., 95%
Assay Percent Range 95%
Molecular Formula C10H20N2
CAS 4897-50-1
Synonym 1, 4'-Bipiperidine
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.