missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
(4-Piperidinophenyl)methylamine, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
214759-73-6 |
Molecular Formula |
C12H18N2 |
Molecular Weight (g/mol) |
190.29 |
MDL Number |
MFCD06659080 |
InChI Key |
WROBJEHXMLQDQP-UHFFFAOYSA-N |
Synonym |
4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine |
PubChem CID |
2795551 |
IUPAC Name |
(4-piperidin-1-ylphenyl)methanamine |
SMILES |
NCC1=CC=C(C=C1)N1CCCCC1 |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
214759-73-6 |
|
190.29 |
|
WROBJEHXMLQDQP-UHFFFAOYSA-N |
|
2795551 |
|
NCC1=CC=C(C=C1)N1CCCCC1 |
|
C12H18N2 |
|
MFCD06659080 |
|
4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine |
|
(4-piperidin-1-ylphenyl)methanamine |
Specifications
|
214759-73-6 |
|
Amber Glass Bottle |
|
MFCD06659080 |
|
4-piperidinophenyl methylamine, 4-piperidin-1-yl phenyl methanamine, 4-piperidinobenzylamine, 4-piperidin-1-yl-benzylamine, 1-4-piperidin-1-yl phenyl methanamine, 4-piperidin-1yl benzylamine, 4-piperidin-1ylphenyl methylamine, 4-piperidylphenyl methylamine, 4-1-piperidyl phenyl methanamine |
|
NCC1=CC=C(C=C1)N1CCCCC1 |
|
190.29 |
|
190.29 |
|
(4-Piperidinophenyl)methylamine |
|
97% |
|
C12H18N2 |
|
1g |
|
WROBJEHXMLQDQP-UHFFFAOYSA-N |
|
(4-piperidin-1-ylphenyl)methanamine |
|
2795551 |
|
97% |
SDS
missing translation for 'documents'