Please login to your online account to display your discounted pricing

4-(piperidinomethyl)benzoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45501CB

 View more versions of this product

Catalog No. CC45501CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 219.28
Quantity 250mg
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17NO2
Health Hazard 1 Warning
Packaging Amber Glass Bottle
Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
CAS 159691-33-5

Specifications

Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17NO2
Formula Weight 219.28
Accessories

Warning

SDS & Safety Documents