Please login to your online account to display your discounted pricing

4-(piperidinomethyl)benzoic acid, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45501EA

 View more versions of this product

Catalog No. CC45501EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 219.28
Quantity 10g
Assay 97%
Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
Assay Percent Range 97%
Molecular Formula C13H17NO2
CAS 159691-33-5
Health Hazard 1 Warning
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Piperidinomethyl)benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C13H17NO2
Formula Weight 219.28
Accessories

Warning