missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidinobenzoic acid, 97%, Maybridge

Chemical Identifiers

CAS 22090-24-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H15NO2
Formula Weight 205.26
MDL Number MFCD00587602
Chemical Name or Material 4-Piperidinobenzoic acid
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC33901CB SDS Fisher Scientific Company
CC33901CB
Amber glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33901DA SDS Fisher Scientific Company
CC33901DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC33901DE Fisher Scientific Company
CC33901DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

22090-24-0
100.0
C12H15NO2
MFCD00587602
97%
96.5
97%
205.26
4-Piperidinobenzoic acid

Specifications

250mg
96.5
Amber glass bottle
C12H15NO2
MFCD00587602
97%
22090-24-0
100.0
97%
205.26
4-Piperidinobenzoic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning