missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperidinemethanol, TCI America™

Manufacturer:  TCI America P165325G

 View more versions of this product

Catalog No. P165325G



Specifications

Specifications

4-Piperidinemethanol
White-Yellow
25g
C6H13NO
4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol
C1CNCCC1CO
115.176
115.18
≥97.0% (GC,T)
135°C
59°C
6457-49-4
3263
XBXHCBLBYQEYTI-UHFFFAOYSA-N
piperidin-4-ylmethanol
420771
Crystal-Powder at 20°C

Chemical Identifiers

6457-49-4
115.176
4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol
piperidin-4-ylmethanol
C6H13NO
XBXHCBLBYQEYTI-UHFFFAOYSA-N
420771
C1CNCCC1CO
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3729

TSCA : No

Documents