missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Piperidineacetic acid hydrochloride, 95%

$28.84 - $337.73

Chemical Identifiers

CAS 73415-84-6
Molecular Formula C7H14ClNO2
Molecular Weight (g/mol) 179.644
MDL Number MFCD03414187
InChI Key IYDUFJOXYMLYNM-UHFFFAOYSA-N
Synonym 4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl
PubChem CID 15565775
IUPAC Name 2-piperidin-4-ylacetic acid;hydrochloride
SMILES C1CNCCC1CC(=O)O.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64031MD View Documents Alfa Aesar™
H64031MD
250mg Each for $28.84
 
AAH6403103 View Documents Alfa Aesar™
H6403103
1g Each for $85.14
 
AAH6403106 View Documents Alfa Aesar™
H6403106
5g Each for $337.73
 
Specifications

Chemical Identifiers

73415-84-6
179.644
IYDUFJOXYMLYNM-UHFFFAOYSA-N
15565775
C1CNCCC1CC(=O)O.Cl
C7H14ClNO2
MFCD03414187
4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl
2-piperidin-4-ylacetic acid;hydrochloride

Specifications

Yellow
73415-84-6
MFCD03414187
IYDUFJOXYMLYNM-UHFFFAOYSA-N
2-piperidin-4-ylacetic acid;hydrochloride
15565775
Solid
4-Piperidineacetic acid hydrochloride
250mg
C7H14ClNO2
4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl
C1CNCCC1CC(=O)O.Cl
179.644
179.64
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning