missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Piperidineacetic acid hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6403103
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Piperidineacetic acid hydrochloride | |
Yellow | |
MFCD03414187 | |
IYDUFJOXYMLYNM-UHFFFAOYSA-N | |
2-piperidin-4-ylacetic acid;hydrochloride | |
15565775 | |
95% | |
Solid |
73415-84-6 | |
C7H14ClNO2 | |
4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl | |
C1CNCCC1CC(=O)O.Cl | |
179.644 | |
179.64 | |
1 g |
Chemical Identifiers
73415-84-6 | |
179.644 | |
IYDUFJOXYMLYNM-UHFFFAOYSA-N | |
15565775 | |
C1CNCCC1CC(=O)O.Cl |
C7H14ClNO2 | |
MFCD03414187 | |
4-piperidineacetic acid hydrochloride, 2-piperidin-4yl acetic acid hydrochloride, 2-piperidin-4-yl acetic acid hydrochloride, 4-piperidineacetic acid hcl, 4-piperidineacetic acid, hydrochloride, piperidin-4-ylacetic acid hydrochloride, piperidine-4-acetic acid hcl, piperidine-4-acetic acid hydrochloride, 4-piperidine acetic acid hydrochloride, piperidin-4-yl-acetic acid hcl | |
2-piperidin-4-ylacetic acid;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures