missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[4-(Piperidin-1-ylmethyl)phenyl]methanol, ≥95%, Maybridge

Chemical Identifiers

CAS 91271-62-4
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C13H19NO
Formula Weight 205.3
MDL Number MFCD07772863
Chemical Name or Material [4-(Piperidin-1-ylmethyl)phenyl]methanol
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45509DA SDS Fisher Scientific Company
CC45509DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

91271-62-4
100.0
C13H19NO
MFCD07772863
95%
94.5
95%
205.3
[4-(Piperidin-1-ylmethyl)phenyl]methanol

Specifications

1g
94.5
Amber Glass Bottle
C13H19NO
MFCD07772863
95%
91271-62-4
100.0
95%
205.3
[4-(Piperidin-1-ylmethyl)phenyl]methanol

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger