Please login to your online account to display your discounted pricing

4-(piperidin-1-ylmethyl)benzaldehyde, 95%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45504DE

 View more versions of this product

Catalog No. CC45504DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 203.28
Quantity 5g
Assay 95%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
Assay Percent Range 95%
Molecular Formula C13H17NO
CAS 471929-86-9
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H17NO
Formula Weight 203.28