missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(piperidin-1-ylmethyl)benzaldehyde, 95%, Maybridge

Chemical Identifiers

Assay Percent Range 95%
Molecular Formula C13H17NO
Formula Weight 203.28
Chemical Name or Material 4-(Piperidin-1-ylmethyl)benzaldehyde
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45504DE Fisher Scientific Company
CC45504DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45504CB SDS Fisher Scientific Company
CC45504CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45504DA SDS Fisher Scientific Company
CC45504DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95%
203.28
95%
C13H17NO
4-(Piperidin-1-ylmethyl)benzaldehyde

Specifications

203.28
471929-86-9
95%
4-(Piperidin-1-ylmethyl)benzaldehyde
5g
Amber Glass Bottle
C13H17NO
95%