Please login to your online account to display your discounted pricing

4-(piperidin-1-ylmethyl)aniline, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-(Piperidin-1-ylmethyl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2
Formula Weight 190.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC45514DA

  SDS
fisher scientific company
CC45514DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.29
Quantity 1g
Assay 97%
CAS 29608-05-7
Assay Percent Range 97%
Molecular Formula C12H18N2
Packaging Amber Glass Bottle
Chemical Name or Material 4-(Piperidin-1-ylmethyl)aniline