Please login to your online account to display your discounted pricing

4-(piperidin-1-ylmethyl)aniline, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC45514EA

 View more versions of this product

Catalog No. CC45514EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.29
Quantity 10g
Assay 97%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)aniline
Molecular Formula C12H18N2
CAS 29608-05-7
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 4-(Piperidin-1-ylmethyl)aniline
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H18N2
Formula Weight 190.29