missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperazinophenol dihydrobromide, 97%, ACROS Organics™

Chemical Identifiers

CAS 38869-37-3
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10 H14 N2 O . 2 Br H
Formula Weight 340.06
MDL Number MFCD00040816
Chemical Name or Material 1-(4-Hydroxyphenyl)piperazine dihydrobromide
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC100330050 SDS Thermo Fisher Scientific
10033-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

38869-37-3
100.0
C10 H14 N2 O . 2 Br H
MFCD00040816
≥97.5%
97.5
98%
340.06
1-(4-Hydroxyphenyl)piperazine dihydrobromide

Specifications

Crystalline Powder
5g
100.0
MFCD00040816
Glass bottle
98%
1-(4-Hydroxyphenyl)piperazine dihydrobromide
340.06
Off-white to light beige or light pink
97.5
Authentic
C10 H14 N2 O . 2 Br H
38869-37-3
≥97.5%

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning