missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperazinoacetophenone, 94%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 159571000

 View more versions of this product

Catalog No. AC159571000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Piperazinoacetophenone
Glass bottle
107°C to 110°C
94%
92.0
94%
MFCD00005962
KPXVKKBJROCIJB-UHFFFAOYSA-N
1-(4-piperazin-1-ylphenyl)ethanone
97269
Powder or Chunks
Authentic
Yellow
100g
51639-48-6
100.0
C12H16N2O
1-4-acetylphenyl piperazine, 1-4-piperazin-1-yl phenyl ethanone, 1-4-piperazin-1-yl-phenyl ethanone, 1-4-piperazin-1-yl-phenyl-ethanone, 1-4-piperazin-1-ylphenyl ethanone, 4'-piperazinoacetophenone, 4-piperazinoacetophenone, p-piperazinoacetophenone, piperazin-4-ylacetophenone
CC(=O)C1=CC=C(C=C1)N2CCNCC2
204.27
204.27
94%

Chemical Identifiers

51639-48-6
204.27
KPXVKKBJROCIJB-UHFFFAOYSA-N
97269
CC(=O)C1=CC=C(C=C1)N2CCNCC2
C12H16N2O
MFCD00005962
1-4-acetylphenyl piperazine, 1-4-piperazin-1-yl phenyl ethanone, 1-4-piperazin-1-yl-phenyl ethanone, 1-4-piperazin-1-yl-phenyl-ethanone, 1-4-piperazin-1-ylphenyl ethanone, 4'-piperazinoacetophenone, 4-piperazinoacetophenone, p-piperazinoacetophenone, piperazin-4-ylacetophenone
1-(4-piperazin-1-ylphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 257-332-6

SDS Documents