missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Piperazinoacetophenone, 94%, Acros Organics

Chemical Identifiers

CAS 51639-48-6
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.27
MDL Number MFCD00005962
InChI Key KPXVKKBJROCIJB-UHFFFAOYSA-N
Synonym 1-4-acetylphenyl piperazine
PubChem CID 97269
IUPAC Name 1-(4-piperazin-1-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCNCC2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC159570050 SDS View Documents Thermo Fisher Scientific
159570050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC159570250 SDS View Documents Thermo Fisher Scientific
159570250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC159571000 SDS View Documents Thermo Fisher Scientific
159571000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

51639-48-6
204.27
KPXVKKBJROCIJB-UHFFFAOYSA-N
97269
CC(=O)C1=CC=C(C=C1)N2CCNCC2
C12H16N2O
MFCD00005962
1-4-acetylphenyl piperazine
1-(4-piperazin-1-ylphenyl)ethanone

Specifications

Authentic
Yellow
5g
92.0
94%
MFCD00005962
KPXVKKBJROCIJB-UHFFFAOYSA-N
1-(4-piperazin-1-ylphenyl)ethanone
97269
Powder or Chunks
4-Piperazinoacetophenone, 94%
Glass bottle
107°C to 110°C
51639-48-6
100.0
C12H16N2O
1-4-acetylphenyl piperazine
CC(=O)C1=CC=C(C=C1)N2CCNCC2
204.27
204.27
94%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 257-332-6