Please login to your online account to display your discounted pricing

4-Piperazin-1-yl-benzonitrile, ≥97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Piperazin-1-yl-benzonitrile
CAS 68104-63-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H13N3
Formula Weight 187.24
MDL Number MFCD01320890
Synonym 4-Piperazin-1-yl-benzonitrile (IUPAC)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07058DA

  SDS
fisher scientific company
MO07058DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO07058EA

  SDS
fisher scientific company
MO07058EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MO07058EB

 
fisher scientific company
MO07058EB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.24
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 97%
Synonym 4-Piperazin-1-yl-benzonitrile (IUPAC)
Molecular Formula C11H13N3
Chemical Name or Material 4-Piperazin-1-yl-benzonitrile
MDL Number MFCD01320890
CAS 68104-63-2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.