Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Picoline N-oxide, 98%

$142.39 - $142.39

Chemical Identifiers

Chemical Name or Material 4-Picoline N-oxide
CAS 1003-67-4
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006210
Beilstein 106329
Synonym 4-Methylpyridine N-oxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1733022

  SDS
alfa aesar™
A1733022
100g Each for $142.39

AAA1733014

  SDS
alfa aesar™
A1733014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Picoline N-oxide
Flash Point 149°C (300°F)
Boiling Point 152°C (11mmHg)
Melting Point 182° to 185°C
Quantity 100g
CAS 1003-67-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UT5800000
TSCA TSCA
Assay 98%
Molecular Formula C6H7NO
Formula Weight 109.13
MDL Number MFCD00006210
Beilstein 106329
Synonym 4-Methylpyridine N-oxide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.