Learn More
4-Picoline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157600010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Picoline | |
98.5 | |
2.4°C | |
0.9500g/mL | |
39°C | |
98.5% min. (GC) | |
C6H7N | |
MFCD00006440 | |
20, 240 | |
15, 7514 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
CC1=CC=NC=C1 | |
93.13 | |
CHEBI:32547 | |
99% |
108-89-4 | |
100.0 | |
Yellow | |
145°C | |
Authentic | |
Glass bottle | |
1.5040 to 1.506 | |
1 L | |
0.95 | |
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy | |
FKNQCJSGGFJEIZ-UHFFFAOYSA-N | |
4-methylpyridine | |
7963 | |
93.13 | |
Liquid |
Chemical Identifiers
108-89-4 | |
93.13 | |
FKNQCJSGGFJEIZ-UHFFFAOYSA-N | |
7963 | |
4-methylpyridine |
C6H7N | |
MFCD00006440 | |
4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy | |
CHEBI:32547 | |
CC1=CC=NC=C1 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide f
GHS Signal Word: Danger
EINECSNumber : 203-626-4
RUO – Research Use Only