Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Phthalimidobutyl)triphenylphosphonium bromide, 97%

$86.66 - $918.95

Chemical Identifiers

Chemical Name or Material (4-Phthalimidobutyl)triphenylphosphonium bromide
CAS 65273-47-4
Molecular Formula C30H27BrNO2P
Formula Weight 544.43
MDL Number MFCD02683092
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2242106

 
alfa aesar™
B2242106
5g Each for $86.66

AAB2242122

 
alfa aesar™
B2242122
100g Each for $918.95
Description & Specifications

Specifications

Formula Weight 544.43
Chemical Name or Material (4-Phthalimidobutyl)triphenylphosphonium bromide
Assay 97%
Molecular Formula C30H27BrNO2P
MDL Number MFCD02683092
CAS 65273-47-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 200° to 202°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.