Please login to your online account to display your discounted pricing

4-Phenyltoluene 98%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  130980050

 View more versions of this product

Catalog No. AC130980050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 168.24
Quantity 5g
Color White to yellow
Beilstein 05, 597
EINECS Number 211-409-0
TSCA TSCA
RTECS Number DV5460000
Melting Point 45° to 49°C
CAS Max % 100.0
CAS Min % 97.5
Chemical Name or Material 4-Phenyltoluene
Name Note 98%
Assay Percent Range 98%
Molecular Formula C13H12
Linear Formula C6H5C6H4CH3
MDL Number MFCD00008544
CAS 644-08-6
Synonym 4-Methylbiphenyl
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Phenyltoluene
Name Note 98%
CAS 644-08-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12
Linear Formula C6H5C6H4CH3
Formula Weight 168.24
MDL Number MFCD00008544
Synonym 4-Methylbiphenyl