Please login to your online account to display your discounted pricing

4-Phenyltoluene 98%, Acros Organics

$138.58 - $138.58

Chemical Identifiers

Chemical Name or Material 4-Phenyltoluene
Name Note 98%
CAS 644-08-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12
Linear Formula C6H5C6H4CH3
Formula Weight 168.24
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC130980050

  SDS
thermo fisher scientific
130980050
5g Each for $138.58
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 168.24
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color White to yellow
Beilstein 05, 597
TSCA TSCA
RTECS Number DV5460000
EINECS Number 211-409-0
Melting Point 45° to 49°C
CAS Max % 100.0
CAS Min % 97.5
Infrared Spectrum Authentic
Synonym 4-Methylbiphenyl
MDL Number MFCD00008544
Linear Formula C6H5C6H4CH3
Chemical Name or Material 4-Phenyltoluene
Name Note 98%
Molecular Formula C13H12
CAS 644-08-6
Assay Percent Range 98%