missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenylthiomorpholine 1,1-Dioxide, TCI America™

Manufacturer:  TCI America P160325G

 View more versions of this product

Catalog No. P160325G



Specifications

Specifications

4-Phenylthiomorpholine 1,1-Dioxide
120°C
17688-68-5
OLQHDKQJHKZUNN-UHFFFAOYSA-N
4-phenyl-1,4-thiazinane 1,1-dioxide
87241
Crystal-Powder at 20°C
Purple-White
25g
C10H13NO2S
C1CS(=O)(=O)CCN1C2=CC=CC=C2
211.279
211.28
≥98.0% (GC)

Chemical Identifiers

17688-68-5
211.279
87241
C1CS(=O)(=O)CCN1C2=CC=CC=C2
C10H13NO2S
OLQHDKQJHKZUNN-UHFFFAOYSA-N
4-phenyl-1,4-thiazinane 1,1-dioxide
Safety and Handling

Safety and Handling

RTECSNumber : XM3050025

TSCA : Yes

Documents