Please login to your online account to display your discounted pricing

(4-phenyltetrahydropyran-4-yl)methylamine, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC69113CB

 View more versions of this product

Catalog No. CC69113CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.27
Quantity 250mg
Assay 95%
Chemical Name or Material (4-phenyltetrahydropyran-4-yl)methylamine
Assay Percent Range 95%
Molecular Formula C12H17NO
CAS 14006-32-7
Health Hazard 1 GHS Signal Word: Danger
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material (4-phenyltetrahydropyran-4-yl)methylamine
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H17NO
Formula Weight 191.27
Accessories

GHS Signal Word: Danger