Please login to your online account to display your discounted pricing

4-Phenyltetrahydropyran-4-carboxylic acid, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC69101DE

 View more versions of this product

Catalog No. CC69101DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 206.24
Quantity 5g
Assay 97%
Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxylic acid
Assay Percent Range 97%
Molecular Formula C12H14O3
CAS 182491-21-0
Health Hazard 1 Warning
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H14O3
Formula Weight 206.24
Accessories

Warning