Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylsemicarbazide, 98+%

$53.73 - $218.58

Chemical Identifiers

Chemical Name or Material 4-Phenylsemicarbazide
CAS 537-47-3
Molecular Formula C6H5NHCONHNH2
Formula Weight 151.17
MDL Number MFCD00007590
Beilstein 511107
Merck Index 14,7312
Synonym N-Phenylhydrazinecarboxamide; Anilinoformylhydrazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2557809

 
alfa aesar™
B2557809
10g Each for $53.73

AAB2557814

 
alfa aesar™
B2557814
25g Each for $114.36

AAB2557818

 
alfa aesar™
B2557818
50g Each for $218.58
Description & Specifications

Specifications

Formula Weight 151.17
Chemical Name or Material 4-Phenylsemicarbazide
Assay ≥98%
Molecular Formula C6H5NHCONHNH2
MDL Number MFCD00007590
CAS 537-47-3
Beilstein 511107
Merck Index 14,7312
Synonym N-Phenylhydrazinecarboxamide; Anilinoformylhydrazine
Melting Point 123° to 125°C
Quantity 10g