Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylquinuclidine, 98%

$346.34 - $346.34

Chemical Identifiers

Chemical Name or Material 4-Phenylquinuclidine
CAS 51069-11-5
Molecular Formula C13H17N
Formula Weight 187.29
MDL Number MFCD00797340
Synonym 4-Phenyl-1-azabicyclo[2.2.2]octane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1552303

 
alfa aesar™
L1552303
1g Each for $346.34
Description & Specifications

Specifications

Formula Weight 187.29
Chemical Name or Material 4-Phenylquinuclidine
Assay 98%
Molecular Formula C13H17N
MDL Number MFCD00797340
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 51069-11-5
Synonym 4-Phenyl-1-azabicyclo[2.2.2]octane
Melting Point 123° to 124°C
Quantity 1g

Applications
4-Phenylquinuclidine is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.