Learn More
4-Phenylphenol, 97%, Thermo Scientific Chemicals
$316.93 - $1053.29
Chemical Identifiers
CAS | 92-69-3 |
---|---|
Molecular Formula | C12H10O |
Molecular Weight (g/mol) | 170.211 |
MDL Number | MFCD00002347 |
InChI Key | YXVFYQXJAXKLAK-UHFFFAOYSA-N |
Synonym | 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol |
PubChem CID | 7103 |
ChEBI | CHEBI:34422 |
IUPAC Name | 4-phenylphenol |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1081736
|
Thermo Scientific Chemicals
A1081736 |
500 g |
Each for $316.93
|
|
|||||
AAA108170E
|
Thermo Scientific Chemicals
A108170E |
2500 g |
Each for $1,053.29
|
|
|||||
Description
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.
Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
92-69-3 | |
170.211 | |
YXVFYQXJAXKLAK-UHFFFAOYSA-N | |
7103 | |
4-phenylphenol |
C12H10O | |
MFCD00002347 | |
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol | |
CHEBI:34422 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)O |
Specifications
92-69-3 | |
305°C to 308°C | |
C12H10O | |
500 g | |
1907452 | |
4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol | |
YXVFYQXJAXKLAK-UHFFFAOYSA-N | |
4-phenylphenol | |
7103 | |
170.21 | |
4-Phenylphenol |
164°C to 168°C | |
165°C (329°F) | |
MFCD00002347 | |
UN3077 | |
14,7305 | |
Soluble in water (0.7mg/ml at 20°C),ether,most common organic solvents,methanol,and chloroform. | |
C1=CC=C(C=C1)C2=CC=C(C=C2)O | |
170.211 | |
CHEBI:34422 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-179-2
RTECSNumber : DV5850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only