Please login to your online account to display your discounted pricing

4-Phenylimidazole, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  163140050

 View more versions of this product

Catalog No. AC163140050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to beige
Chemical Name or Material 4-Phenylimidazole
Name Note 99%
CAS 670-95-1
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to beige granular powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8N2
Formula Weight 144.18g/mol
MDL Number MFCD00005197
Beilstein 23, 182
Health Hazard 1 Warning
EINECS Number 211-580-1

Specifications

Chemical Name or Material 4-Phenylimidazole
Name Note 99%
CAS 670-95-1
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8N2
Formula Weight 144.18g/mol
MDL Number MFCD00005197
Accessories

Warning

SDS & Safety Documents