Please login to your online account to display your discounted pricing

4-Phenylimidazole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Phenylimidazole
Name Note 99%
CAS 670-95-1
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8N2
Formula Weight 144.18g/mol
MDL Number MFCD00005197
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163140050

  SDS
fisher scientific acros
163140050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color White to beige
Chemical Name or Material 4-Phenylimidazole
Name Note 99%
CAS 670-95-1
CAS Min % 98.5
CAS Max % 100.0
Physical Form White to beige granular powder
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C9H8N2
Formula Weight 144.18g/mol
MDL Number MFCD00005197
Beilstein 23, 182
Health Hazard 1 Warning
EINECS Number 211-580-1
Safety and Handling

Warning