missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Phenylethynyl)benzeneboronic acid pinacol ester, 97%

$96.93 - $388.95

Chemical Identifiers

CAS 1190376-20-5
Molecular Formula C20H21BO2
Formula Weight 304.19
MDL Number MFCD16294498
Beilstein 20037873
Synonym Diphenylacetylene-4-boronic acid pinacol ester; 4,4,5,5-Tetramethyl-2-(4-phenylethynyl)-1,3,2-dioxaborolane
Chemical Name or Material 4-(Phenylethynyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5169903 SDS Alfa Aesar™
H5169903
1g Each for $96.93
AAH5169906 SDS Alfa Aesar™
H5169906
5g Each for $388.95
Specifications Description & Specifications

Chemical Identifiers

1190376-20-5
304.19
20037873
4-(Phenylethynyl)benzeneboronic acid pinacol ester
C20H21BO2
MFCD16294498
Diphenylacetylene-4-boronic acid pinacol ester; 4,4,5,5-Tetramethyl-2-(4-phenylethynyl)-1,3,2-dioxaborolane

Specifications

1g
C20H21BO2
MFCD16294498
Diphenylacetylene-4-boronic acid pinacol ester; 4,4,5,5-Tetramethyl-2-(4-phenylethynyl)-1,3,2-dioxaborolane
97%
1190376-20-5
304.19
20037873
4-(Phenylethynyl)benzeneboronic acid pinacol ester