Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Phenylethynyl)acetophenone, 97%

$56.50 - $200.41

Chemical Identifiers

Chemical Name or Material 4'-(Phenylethynyl)acetophenone
CAS 1942-31-0
Molecular Formula C16H12O
Formula Weight 220.27
MDL Number MFCD00219774
Synonym 4-Acetyldiphenylacetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1937003

  SDS
alfa aesar™
A1937003
1g Each for $56.50

AAA1937006

  SDS
alfa aesar™
A1937006
5g Each for $200.41
Description & Specifications

Specifications

Formula Weight 220.27
Chemical Name or Material 4'-(Phenylethynyl)acetophenone
Assay 97%
Molecular Formula C16H12O
MDL Number MFCD00219774
CAS 1942-31-0
Synonym 4-Acetyldiphenylacetylene
Melting Point 99° to 102°C
Quantity 1g

Applications
Fluorescence experiments on 4′-phenylethynyl-acetophenone, whose fluorescence quantum yield was found to be less than 0.001. Reactant in the synthesis of trifluoromethyltrimethylsilane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.