Learn More
4-Phenylcinnamic acid, 98%, Thermo Scientific Chemicals
$226.64 - $911.88
Chemical Identifiers
CAS | 13026-23-8 |
---|---|
Molecular Formula | C15H12O2 |
Molecular Weight (g/mol) | 224.26 |
MDL Number | MFCD00014010 |
InChI Key | DMJDEZUEYXVYNO-FLIBITNWSA-N |
Synonym | 4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid |
PubChem CID | 5842785 |
IUPAC Name | (E)-3-(4-phenylphenyl)prop-2-enoic acid |
SMILES | OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1264309
|
Thermo Scientific Chemicals
A1264309 |
10 g |
Each for $226.64
|
|
|||||
AAA1264314
|
Thermo Scientific Chemicals
A1264314 |
25 g |
Each for $514.78
|
|
|||||
AAA1264318
|
Thermo Scientific Chemicals
A1264318 |
50 g |
Each for $911.88
|
|
|||||
Description
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Chemical Identifiers
13026-23-8 | |
224.26 | |
DMJDEZUEYXVYNO-FLIBITNWSA-N | |
5842785 | |
OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1 |
C15H12O2 | |
MFCD00014010 | |
4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid | |
(E)-3-(4-phenylphenyl)prop-2-enoic acid |
Specifications
13026-23-8 | |
C15H12O2 | |
10 g | |
4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid | |
DMJDEZUEYXVYNO-FLIBITNWSA-N | |
(E)-3-(4-phenylphenyl)prop-2-enoic acid | |
5842785 | |
98% |
224°C to 228°C | |
MFCD00014010 | |
1955124 | |
Slightly soluble in water. | |
OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1 | |
224.26 | |
224.27 | |
4-Phenylcinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-888-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only