missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenylcinnamic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1264314

 View more versions of this product

Catalog No. AAA1264314



Description

Description

4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

4-Phenylcinnamic acid
224°C to 228°C
MFCD00014010
1955124
Slightly soluble in water.
OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1
224.26
224.27
13026-23-8
C15H12O2
25 g
4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid
DMJDEZUEYXVYNO-FLIBITNWSA-N
(E)-3-(4-phenylphenyl)prop-2-enoic acid
5842785
98%

Chemical Identifiers

13026-23-8
224.26
DMJDEZUEYXVYNO-FLIBITNWSA-N
5842785
OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1
C15H12O2
MFCD00014010
4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid
(E)-3-(4-phenylphenyl)prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 235-888-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only