Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylchalcone Oxide, 99%


Quantity 20mg

Manufacturer: alfa aesar™  J67334FPL

 View more versions of this product

Catalog No. AAJ67334FPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 257° to 259°C
Formula Weight 300.36
Chemical Name or Material 4-Phenylchalcone Oxide
MDL Number MFCD00211155
Synonym 4-PCO
CAS 82389-34-2
Physical Form Solid
Assay 99%
Quantity 20mg
Storage Note 1 -30° to -10°C
Color White

Specifications

Chemical Name or Material 4-Phenylchalcone Oxide
CAS 82389-34-2
Formula Weight 300.36
MDL Number MFCD00211155
Synonym 4-PCO
Accessories

-30° to -10°C