Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Phenylchalcone Oxide, 99%

Chemical Identifiers

Chemical Name or Material 4-Phenylchalcone Oxide
CAS 82389-34-2
Formula Weight 300.36
MDL Number MFCD00211155
Synonym 4-PCO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67334FPL

 
alfa aesar™
J67334FPL
20mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 257° to 259°C
Formula Weight 300.36
Chemical Name or Material 4-Phenylchalcone Oxide
MDL Number MFCD00211155
Synonym 4-PCO
CAS 82389-34-2
Physical Form Solid
Assay 99%
Quantity 20mg
Storage Note 1 -30° to -10°C
Color White
Safety and Handling

-30° to -10°C